ENA181911603 Enamine

N-[4-(methylsulfonyl)phenyl]urea

  • MDL number MFCD09932960
  • Cas Number : 343247-75-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 224...226
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-37210

Safety Information

msds https://www.enaminestore.com/catalog/EN300-37210
Availability details for
Quantity
  • Availability
  • Add to Cart