ENA181911611 Enamine

N-benzyl-2-hydroxyacetamide

  • MDL number MFCD08443475
  • Cas Number : 19340-77-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 103-104
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-37230

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-37230
Availability details for
Quantity
  • Availability
  • Add to Cart