ENA181911614 Enamine

2-tert-butyl-4-(chloromethyl)-1,3-thiazole

  • MDL number MFCD09693922
  • Cas Number : 675149-83-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-37233

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-37233
Availability details for
Quantity
  • Availability
  • Add to Cart