ENA181911618 Enamine

tert-butyl N-[(3-aminophenyl)methyl]-N-methylcarbamate

  • MDL number MFCD11099516
  • Cas Number : 167756-90-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-37241

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-37241
Availability details for
Quantity
  • Availability
  • Add to Cart