ENA181911621 Enamine

1-(4-fluorophenyl)-3-methyl-1H-pyrazolo[3,4-b]pyridine-5-carboxylic acid

  • MDL number MFCD09042636
  • Cas Number : 926193-95-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-37244

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-37244
Availability details for
Quantity
  • Availability
  • Add to Cart