ENA181911626 Enamine

4-(5-aminopyridin-2-yl)piperazin-2-one

  • MDL number MFCD09040668
  • Cas Number : 926262-86-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 136...138
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-37254

Safety Information

msds https://www.enaminestore.com/catalog/EN300-37254
Availability details for
Quantity
  • Availability
  • Add to Cart