ENA181911631 Enamine

N-(3-cyanophenyl)-2-fluorobenzamide

  • MDL number MFCD04070710
  • Cas Number : 709647-30-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 132...134
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-37260

Safety Information

msds https://www.enaminestore.com/catalog/EN300-37260
Availability details for
Quantity
  • Availability
  • Add to Cart