ENA181911633 Enamine

N-(1-benzylpiperidin-4-yl)-3-fluorobenzamide

  • MDL number MFCD01480540
  • Cas Number : 945119-01-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-37263

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-37263
Availability details for
Quantity
  • Availability
  • Add to Cart