ENA181911641 Enamine

5-(propan-2-yl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD08443863
  • Cas Number : 870704-28-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-37272

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-37272
Availability details for
Quantity
  • Availability
  • Add to Cart