ENA181911656 Enamine

4-methoxy-3-methylbenzenesulfonyl chloride

  • MDL number MFCD08445775
  • Cas Number : 84910-98-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 51...53
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-37290

Safety Information

msds https://www.enaminestore.com/catalog/EN300-37290
Availability details for
Quantity
  • Availability
  • Add to Cart