ENA181911658 Enamine

2-(1-methyl-1H-pyrazole-4-sulfonamido)acetic acid

  • MDL number MFCD11099526
  • Cas Number : 1152914-93-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-37292

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-37292
Availability details for
Quantity
  • Availability
  • Add to Cart