ENA181911660 Enamine

2-(3-aminophenoxy)-1-(pyrrolidin-1-yl)ethan-1-one

  • MDL number MFCD09049724
  • Cas Number : 926242-86-6
  • Molecular Weight : 220.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-37296

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-37296
Availability details for
Quantity
  • Availability
  • Add to Cart