ENA181911662 Enamine

N-(3-amino-4-chlorophenyl)methanesulfonamide

  • MDL number MFCD09050053
  • Cas Number : 77947-01-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 156...158
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-37298

Safety Information

msds https://www.enaminestore.com/catalog/EN300-37298
Availability details for
Quantity
  • Availability
  • Add to Cart