ENA181911672 Enamine

4-phenyloxane-4-carbonitrile

  • MDL number MFCD00085697
  • Cas Number : 1202-81-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-37310

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-37310
Availability details for
Quantity
  • Availability
  • Add to Cart