ENA181911684 Enamine

N-methyl-N-(2,2,2-trifluoroethyl)amine hydrochloride

  • MDL number MFCD08448126
  • Cas Number : 2730-52-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 195...197
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-37329

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-37329
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart