ENA181911685 Enamine

tert-butyl 3-aminopropyl(methyl)carbamate

  • MDL number MFCD01631189
  • Cas Number : 150349-36-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-37330

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-37330
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart