ENA181911689 Enamine

(6-fluoro-4H-1,3-benzodioxin-8-yl)acetonitrile

  • MDL number MFCD11099533
  • Cas Number : 1087784-38-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 75...77
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-37334

Safety Information

msds https://www.enaminestore.com/catalog/EN300-37334
Availability details for
Quantity
  • Availability
  • Add to Cart