ENA181911691 Enamine

4-hydrazinyl-1-methyl-1H-pyrazolo[3,4-d]pyrimidine

  • MDL number MFCD04051940
  • Cas Number : 5334-72-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-37337

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-37337
Availability details for
Quantity
  • Availability
  • Add to Cart