ENA181911692 Enamine

2-[4-(4-fluorophenyl)piperazin-1-yl]ethanamine

  • MDL number MFCD09723204
  • Cas Number : 27144-84-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-37340

Safety Information

msds https://www.enaminestore.com/catalog/EN300-37340
Availability details for
Quantity
  • Availability
  • Add to Cart