ENA181911694 Enamine

2-(3-aminophenoxy)-N-ethylacetamide

  • MDL number MFCD09047521
  • Cas Number : 926221-29-6
  • Molecular Weight : 194.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-37342

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-37342
Availability details for
Quantity
  • Availability
  • Add to Cart