ENA181911698 Enamine

N-[1-(2-tert-butyl-4-methyl-1,3-thiazol-5-yl)ethylidene]hydroxylamine

  • Cas Number : 1087792-29-7
  • Molecular Weight : 212.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-37346

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-37346
Availability details for
Quantity
  • Availability
  • Add to Cart