ENA181911699 Enamine

2-(2-benzyl-1,3-thiazol-4-yl)acetic acid

  • MDL number MFCD10007085
  • Cas Number : 858486-04-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 126-128
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-37347

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-37347
Availability details for
Quantity
  • Availability
  • Add to Cart