ENA181911705 Enamine

1-(4-chloro-3-nitrophenyl)pyrrolidin-2-one

  • MDL number MFCD11099539
  • Cas Number : 1087784-40-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-37356

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-37356
Availability details for
Quantity
  • Availability
  • Add to Cart