ENA181911710 Enamine

2-[5-(dimethylsulfamoyl)-2-methoxyphenyl]acetic acid

  • MDL number MFCD09042972
  • Cas Number : 926249-45-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-37362

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-37362
Availability details for
Quantity
  • Availability
  • Add to Cart