ENA181911715 Enamine

2-(naphthalen-1-yloxy)-1-(piperazin-1-yl)ethan-1-one

  • MDL number MFCD04116573
  • Cas Number : 610802-14-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-37372

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-37372
Availability details for
Quantity
  • Availability
  • Add to Cart