ENA181911722 Enamine

3-[(5-aminopyridin-2-yl)oxy]benzonitrile

  • MDL number MFCD09810305
  • Cas Number : 1016705-78-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-37380

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-37380
Availability details for
Quantity
  • Availability
  • Add to Cart