ENA181911724 Enamine

3-cyano-N-[(furan-2-yl)methyl]benzamide

  • MDL number MFCD08495254
  • Cas Number : 879183-68-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-37382

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-37382
Availability details for
Quantity
  • Availability
  • Add to Cart