ENA181911727 Enamine

5-bromo-3-chloro-1-methyl-1H-indole-2-carboxylic acid

  • MDL number MFCD11052582
  • Cas Number : 1057404-45-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-37386

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-37386
Availability details for
Quantity
  • Availability
  • Add to Cart