ENA181911730 Enamine

2-(piperazine-1-carbonyl)phenol hydrochloride

  • MDL number MFCD11099545
  • Cas Number : 1170899-10-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-37389

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-37389
Availability details for
Quantity
  • Availability
  • Add to Cart