ENA181911736 Enamine

2,2-dimethylmorpholine

  • MDL number MFCD09701959
  • Cas Number : 147688-58-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-37404

Safety Information

signalword Danger
pcodes P210,P233,P240,P241,P242,P243,P280,P303+P361+P353,P305+P351+P338,P310,P370+P378,P403+P235,P501
hcodes Xi
MSDS https://www.enaminestore.com/catalog/EN300-37404
pictograms GHS02,GHS05
hstatements H226,H318
Availability details for
Quantity
  • Availability
  • Add to Cart