ENA181911739 Enamine

2-oxo-3-(thiophen-2-yl)propanoic acid

  • MDL number MFCD04971657
  • Cas Number : 15504-41-3
  • Molecular Weight : 170.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-37407

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-37407
Availability details for
Quantity
  • Availability
  • Add to Cart