ENA181911748 Enamine

4-(2,6-difluorophenyl)-1,3-thiazol-2-amine

  • MDL number MFCD03713937
  • Cas Number : 510738-34-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 180-181
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-37417

Safety Information

signalword Warning
pcodes P264,P270,P301+P312,P330,P501
hcodes Xn
MSDS https://www.enaminestore.com/catalog/EN300-37417
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart