ENA181911754 Enamine

6-methyl-2-(3-nitrophenyl)pyrimidin-1-ium-1-olate

  • MDL number MFCD11099550
  • Cas Number : 210036-45-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-37425

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-37425
Availability details for
Quantity
  • Availability
  • Add to Cart