ENA181911762 Enamine

2-(benzyloxy)propanoic acid

  • MDL number MFCD02093932
  • Cas Number : 6625-78-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 51...53
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-37443

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-37443
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart