ENA181911765 Enamine

3-(4-ethylpiperazin-1-yl)propanenitrile

  • MDL number MFCD09928036
  • Cas Number : 4491-93-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-37451

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-37451
Availability details for
Quantity
  • Availability
  • Add to Cart