ENA181911776 Enamine

3-(5-methyl-2-nitrophenoxy)propanoic acid

  • MDL number MFCD09948263
  • Cas Number : 1016865-66-9
  • Molecular Weight : 225.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-37475

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-37475
Availability details for
Quantity
  • Availability
  • Add to Cart