ENA181911780 Enamine

methyl 3-hydroxy-2-[(triphenylmethyl)amino]butanoate

  • MDL number MFCD08517754
  • Cas Number : 74481-55-3
  • Molecular Weight : 375.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-37481

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-37481
Availability details for
Quantity
  • Availability
  • Add to Cart