ENA181911785 Enamine

4-(3-nitrobenzenesulfonyl)piperazin-2-one

  • MDL number MFCD11099556
  • Cas Number : 1087784-42-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-37491

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-37491
Availability details for
Quantity
  • Availability
  • Add to Cart