ENA181911794 Enamine

N-(2-methylphenyl)hydrazinecarboxamide

  • MDL number MFCD00152881
  • Cas Number : 15940-63-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 141...143
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-37515

Safety Information

msds https://www.enaminestore.com/catalog/EN300-37515
Availability details for
Quantity
  • Availability
  • Add to Cart