ENA181911795 Enamine

{2-[(thien-2-ylacetyl)amino]-1,3-thiazol-4-yl}acetic acid

  • MDL number MFCD02043420
  • Cas Number : 332099-91-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 200...202
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-37516

Safety Information

msds https://www.enaminestore.com/catalog/EN300-37516
Availability details for
Quantity
  • Availability
  • Add to Cart