ENA181911796 Enamine

(2Z)-3-(4-aminophenyl)-2-cyanoacrylamide

  • MDL number MFCD11099559
  • Cas Number : 30980-78-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-37517

Safety Information

msds https://www.enaminestore.com/catalog/EN300-37517
Availability details for
Quantity
  • Availability
  • Add to Cart