ENA181911804 Enamine

1-(2-nitrophenyl)-1H-1,2,3,4-tetrazole

  • MDL number MFCD00457932
  • Cas Number : 14210-50-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-37527

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-37527
Availability details for
Quantity
  • Availability
  • Add to Cart