ENA181911807 Enamine

N-(5-methylisoxazol-3-yl)acrylamide

  • MDL number MFCD11099378
  • Cas Number : 196403-11-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 169...171
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-22991

Safety Information

msds https://www.enaminestore.com/catalog/EN300-22991
Availability details for
Quantity
  • Availability
  • Add to Cart