ENA181911811 Enamine

N-(1-benzylpiperidin-4-yl)furan-2-carboxamide

  • MDL number MFCD00832302
  • Cas Number : 132864-14-3
  • Molecular Weight : 284.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-37534

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-37534
Availability details for
Quantity
  • Availability
  • Add to Cart