ENA181911815 Enamine

3-(cyclopentylformamido)propanoic acid

  • MDL number MFCD08445014
  • Cas Number : 923225-18-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-37539

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-37539
Availability details for
Quantity
  • Availability
  • Add to Cart