ENA181911822 Enamine

ethyl[(3-methylphenyl)methyl]amine

  • MDL number MFCD00048518
  • Cas Number : 209051-77-4
  • Molecular Weight : 149.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-32109

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-32109
Availability details for
Quantity
  • Availability
  • Add to Cart