ENA181911827 Enamine

5-amino-2-chloro-N,N-dimethylbenzamide

  • MDL number MFCD09741200
  • Cas Number : 946691-01-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 158...160
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-37553

Safety Information

msds https://www.enaminestore.com/catalog/EN300-37553
Availability details for
Quantity
  • Availability
  • Add to Cart