ENA181911839 Enamine

2-[(5-aminopyridin-2-yl)oxy]benzonitrile

  • MDL number MFCD09802361
  • Cas Number : 868697-72-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-37570

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-37570
Availability details for
Quantity
  • Availability
  • Add to Cart