ENA181911840 Enamine

2-(4-methoxyphenoxy)pyridin-3-amine

  • MDL number MFCD01570916
  • Cas Number : 170440-07-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-37571

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-37571
Availability details for
Quantity
  • Availability
  • Add to Cart