ENA181911842 Enamine

1,5-diisopropyl-2,4-dinitrobenzene

  • MDL number MFCD07779257
  • Cas Number : 77256-78-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 77...79
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-37574

Safety Information

msds https://www.enaminestore.com/catalog/EN300-37574
Availability details for
Quantity
  • Availability
  • Add to Cart